Learn More
4,4'-Methylenebis(N-phenylmaleimide), 95%, Thermo Scientific Chemicals
$82.26 - $82.26
Chemical Identifiers
| CAS | 13676-54-5 |
|---|---|
| Molecular Formula | C21H14N2O4 |
| Molecular Weight (g/mol) | 358.353 |
| MDL Number | MFCD00005507 |
| InChI Key | XQUPVDVFXZDTLT-UHFFFAOYSA-N |
| Synonym | bismaleimide, bismaleimide s, bis 4-maleimidophenyl methane, diphenylmethanebismaleimide, p,p'-dimaleimidodiphenylmethane, 4,4'-methylenebis n-phenylmaleimide, bis p-maleimidophenyl methane, 4,4'-dimaleimidodiphenylmethane, 4,4'-diphenylmethanedimaleimide, 4,4'-biphenylmethanebismaleimide |
| PubChem CID | 83648 |
| IUPAC Name | 1-[4-[[4-(2,5-dioxopyrrol-1-yl)phenyl]methyl]phenyl]pyrrole-2,5-dione |
| SMILES | C1=CC(=CC=C1CC2=CC=C(C=C2)N3C(=O)C=CC3=O)N4C(=O)C=CC4=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAL1286214
|
Thermo Scientific Chemicals
L1286214 |
25 g |
Each for $82.26
|
|
|||||
Description
4,4'-Methylenebis(N-phenylmaleimide) was used to cure glycidyl ester of eleostearic acid (GEEA).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4,4′-Methylenebis(N-phenylmaleimide) was used to cure glycidyl ester of eleostearic acid (GEEA).
Solubility
Insoluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Chemical Identifiers
| 13676-54-5 | |
| 358.353 | |
| XQUPVDVFXZDTLT-UHFFFAOYSA-N | |
| 83648 | |
| C1=CC(=CC=C1CC2=CC=C(C=C2)N3C(=O)C=CC3=O)N4C(=O)C=CC4=O |
| C21H14N2O4 | |
| MFCD00005507 | |
| bismaleimide, bismaleimide s, bis 4-maleimidophenyl methane, diphenylmethanebismaleimide, p,p'-dimaleimidodiphenylmethane, 4,4'-methylenebis n-phenylmaleimide, bis p-maleimidophenyl methane, 4,4'-dimaleimidodiphenylmethane, 4,4'-diphenylmethanedimaleimide, 4,4'-biphenylmethanebismaleimide | |
| 1-[4-[[4-(2,5-dioxopyrrol-1-yl)phenyl]methyl]phenyl]pyrrole-2,5-dione |
Specifications
| 13676-54-5 | |
| C21H14N2O4 | |
| 25 g | |
| 333986 | |
| Insoluble in water. | |
| C1=CC(=CC=C1CC2=CC=C(C=C2)N3C(=O)C=CC3=O)N4C(=O)C=CC4=O | |
| 358.353 | |
| 358.36 | |
| 4,4'-Methylenebis(N-phenylmaleimide) |
| 156°C to 158°C | |
| MFCD00005507 | |
| UN2811 | |
| bismaleimide, bismaleimide s, bis 4-maleimidophenyl methane, diphenylmethanebismaleimide, p,p'-dimaleimidodiphenylmethane, 4,4'-methylenebis n-phenylmaleimide, bis p-maleimidophenyl methane, 4,4'-dimaleimidodiphenylmethane, 4,4'-diphenylmethanedimaleimide, 4,4'-biphenylmethanebismaleimide | |
| XQUPVDVFXZDTLT-UHFFFAOYSA-N | |
| 1-[4-[[4-(2,5-dioxopyrrol-1-yl)phenyl]methyl]phenyl]pyrrole-2,5-dione | |
| 83648 | |
| 95% |
Safety and Handling
GHS H Statement
H330-H315-H319-H335
Fatal if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P260-P264b-P271-P280-P284-P302+P352-P304+P340-P305+P351+P338-P310-P312-P332+P313-P362-P501c
H315-H319-H330-H335
DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber : 237-163-4
RTECSNumber : ON5670000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only