missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4,4'-Methylenebis(N,N-diglycidylaniline) 98.0+%, TCI America™
$44.73 - $126.76
Chemical Identifiers
| CAS | 28768-32-3 |
|---|---|
| Molecular Formula | C25H30N2O4 |
| Molecular Weight (g/mol) | 422.525 |
| MDL Number | MFCD00661161 |
| InChI Key | FAUAZXVRLVIARB-UHFFFAOYSA-N |
| PubChem CID | 91593 |
| IUPAC Name | 4-[[4-[bis(oxiran-2-ylmethyl)amino]phenyl]methyl]-N,N-bis(oxiran-2-ylmethyl)aniline |
| SMILES | C1C(O1)CN(CC2CO2)C3=CC=C(C=C3)CC4=CC=C(C=C4)N(CC5CO5)CC6CO6 |
Chemical Identifiers
| 28768-32-3 | |
| 422.525 | |
| FAUAZXVRLVIARB-UHFFFAOYSA-N | |
| 4-[[4-[bis(oxiran-2-ylmethyl)amino]phenyl]methyl]-N,N-bis(oxiran-2-ylmethyl)aniline |
| C25H30N2O4 | |
| MFCD00661161 | |
| 91593 | |
| C1C(O1)CN(CC2CO2)C3=CC=C(C=C3)CC4=CC=C(C=C4)N(CC5CO5)CC6CO6 |
Specifications
| 28768-32-3 | |
| MFCD00661161 | |
| 3082 | |
| C1C(O1)CN(CC2CO2)C3=CC=C(C=C3)CC4=CC=C(C=C4)N(CC5CO5)CC6CO6 | |
| 422.525 | |
| 422.53 | |
| Liquid |
| C25H30N2O4 | |
| 25 g | |
| FAUAZXVRLVIARB-UHFFFAOYSA-N | |
| 4-[[4-[bis(oxiran-2-ylmethyl)amino]phenyl]methyl]-N,N-bis(oxiran-2-ylmethyl)aniline | |
| 91593 | |
| ≥98.0% (T) | |
| 4,4′-Methylenebis(N,N-diglycidylaniline) |
Safety and Handling
TSCA : Yes