missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4,4'-Methylenebis(N,N-diglycidylaniline) 98.0+%, TCI America™
Specifications
Specifications
| Quantity | 25 g |
| UN Number | 3082 |
| Formula Weight | 422.53 |
| Percent Purity | ≥98.0% (T) |
| Physical Form | Liquid |
| Chemical Name or Material | 4,4′-Methylenebis(N,N-diglycidylaniline) |
Safety and Handling
| TSCA | Yes |