missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Methylenebis(2,6-di-tert-butylphenol) 98.0+%, TCI America™

Catalog No. M0214500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M0214500G 500 g
M021425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M0214500G Supplier TCI America Supplier No. M0214500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 118-82-1
Molecular Formula C29H44O2
Molecular Weight (g/mol) 424.669
MDL Number MFCD00008822
InChI Key MDWVSAYEQPLWMX-UHFFFAOYSA-N
Synonym 4,4'-methylenebis 2,6-di-tert-butylphenol, bimox m, ionox 220, binox m, antioxidant e 702, mb 1 antioxidant, ethyl 702, ionox 220 antioxidant, etil 702, 4,4'-methylenebis 2,6-di-t-butylphenol
PubChem CID 8372
ChEBI CHEBI:34369
IUPAC Name 2,6-ditert-butyl-4-[(3,5-ditert-butyl-4-hydroxyphenyl)methyl]phenol
SMILES CC(C)(C)C1=CC(=CC(=C1O)C(C)(C)C)CC2=CC(=C(C(=C2)C(C)(C)C)O)C(C)(C)C

Specifications

Melting Point 155°C
Color White-Yellow
Quantity 500 g
Formula Weight 424.67
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4,4′-Methylenebis(2,6-di-tert-butylphenol)
EINECSNumber (4)-0039
RTECSNumber SL9650000
TSCA Yes