missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Methylenebis(2,6-di-tert-butylphenol), 98%

Catalog No. p-7024370
Change view
Click to view available options
Quantity:
50 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1891018 50 g
AAA1891022 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1891018 Supplier Thermo Scientific Chemicals Supplier No. A1891018
Only null left

CAS: 118-82-1 | C29H44O2 | 424.669 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 118-82-1
Molecular Formula C29H44O2
Molecular Weight (g/mol) 424.669
MDL Number MFCD00008822
InChI Key MDWVSAYEQPLWMX-UHFFFAOYSA-N
Synonym 4,4'-methylenebis 2,6-di-tert-butylphenol, bimox m, ionox 220, binox m, antioxidant e 702, mb 1 antioxidant, ethyl 702, ionox 220 antioxidant, etil 702, 4,4'-methylenebis 2,6-di-t-butylphenol
PubChem CID 8372
ChEBI CHEBI:34369
IUPAC Name 2,6-ditert-butyl-4-[(3,5-ditert-butyl-4-hydroxyphenyl)methyl]phenol
SMILES CC(C)(C)C1=CC(=CC(=C1O)C(C)(C)C)CC2=CC(=C(C(=C2)C(C)(C)C)O)C(C)(C)C

Specifications

Melting Point 154°C to 158°C
Boiling Point 289°C (40 mmHg)
Quantity 50 g
Beilstein 1916919
Formula Weight 424.67
Percent Purity 98%
Chemical Name or Material 4,4'-Methylenebis(2,6-di-tert-butylphenol)
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 204-279-1
RTECSNumber SL9650000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only