missing translation for 'onlineSavingsMsg'
Learn More

4,4-Methylene-bis(2-methylaniline), MilliporeSigma™ Supelco™

Catalog No. 111019786
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111019786 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111019786 Supplier Merck Emd Millipore Supplier No. 46106100MG

4,4′-Methylene-bis(2-methylaniline) belongs to the primary aromatic amines (PAAs) family of compounds which are toxic in nature and are probable human carcinogens.

Cleaning Products, Cosmetics, Environmental, Food and Beverages and Personal Care

Specifications

CAS 838-88-0
Physical Form Neat
Linear Formula C15H18N2
MDL Number MFCD00126963
Synonym 4,4 ′-Diamino-3,3 ′-dimethyldiphenylmethane; 4,4 ′-Methylene-di(o-toluidine)
Type Analytical Standard
Quantity 100 mg
Formula Weight 226.32
Grade Analytical Standard