missing translation for 'onlineSavingsMsg'
Learn More

4-(4-Methoxyphenyl)butyric Acid 95.0+%, TCI America™

Catalog No. p-7132983
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M20375G 5 g
M203725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M20375G Supplier TCI America Supplier No. M20375G
Only null left

Chemical Identifiers

CAS 4521-28-2
Molecular Formula C11H14O3
Molecular Weight (g/mol) 194.23
MDL Number MFCD00004404
InChI Key LZHMNCJMXQKSBY-UHFFFAOYSA-N
Synonym 4-4-methoxyphenyl butyric acid, 4-4-methoxyphenyl butanoic acid, benzenebutanoic acid, 4-methoxy, 4-4-methoxyphenyl butyricacid, 4-4-methoxyphenyl-1-butyric acid, p-methoxy-g-phenylbutyric acid, pubchem15207, 4-methoxybenzenebutyric acid, 4-p-methoxyphenylbutyric acid
PubChem CID 78280
IUPAC Name 4-(4-methoxyphenyl)butanoic acid
SMILES COC1=CC=C(C=C1)CCCC(=O)O

Specifications

Melting Point 60°C
Boiling Point 196°C
Quantity 5 g
Formula Weight 194.23
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-(4-Methoxyphenyl)butyric Acid