missing translation for 'onlineSavingsMsg'
Learn More

4-(4-Methoxyphenyl)butyric acid, 98%

Catalog No. p-7058114
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6061303 1 g
AAH6061306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6061303 Supplier Thermo Scientific Chemicals Supplier No. H6061303

CAS: 4521-28-2 | C11H14O3 | 194.23 g/mol

4-(4-Methoxyphenyl)butyric acid on demethylation with pyridinium hydrochloride affords 4-hydroxyphenylbutyric acid, a key starting material for the synthesis of preclinical candidate LY518674.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-(4-Methoxyphenyl)butyric acid on demethylation with pyridinium hydrochloride affords 4-hydroxyphenylbutyric acid, a key starting material for the synthesis of preclinical candidate LY518674.

Solubility
Slightly soluble in methanol. Insoluble in chloroform.

Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.

Chemical Identifiers

CAS 4521-28-2
Molecular Formula C11H14O3
Molecular Weight (g/mol) 194.23
MDL Number MFCD00004404
InChI Key LZHMNCJMXQKSBY-UHFFFAOYSA-N
Synonym 4-4-methoxyphenyl butyric acid, 4-4-methoxyphenyl butanoic acid, benzenebutanoic acid, 4-methoxy, 4-4-methoxyphenyl butyricacid, 4-4-methoxyphenyl-1-butyric acid, p-methoxy-g-phenylbutyric acid, pubchem15207, 4-methoxybenzenebutyric acid, 4-p-methoxyphenylbutyric acid
PubChem CID 78280
IUPAC Name 4-(4-methoxyphenyl)butanoic acid
SMILES COC1=CC=C(C=C1)CCCC(=O)O

Specifications

Melting Point 56°C to 59°C
Quantity 1 g
Beilstein 2416568
Solubility Information Slightly soluble in methanol. Insoluble in chloroform.
Formula Weight 194.23
Percent Purity 98%
Chemical Name or Material 4-(4-Methoxyphenyl)butyric acid
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 224-849-3
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only