missing translation for 'onlineSavingsMsg'
Learn More

4'-(4-Methoxyphenoxy)acetophenone 98.0+%, TCI America™

Catalog No. p-7133399
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M27401G 1 g
M27405G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M27401G Supplier TCI America Supplier No. M27401G

Chemical Identifiers

CAS 54916-28-8
Molecular Formula C15H14O3
Molecular Weight (g/mol) 242.274
MDL Number MFCD00052928
InChI Key ICXPZMQQZWKKMN-UHFFFAOYSA-N
Synonym 4-acetyl-4'-methoxydiphenyl ether, 1-4-4-methoxyphenoxy phenyl ethanone, 1-4-4-methoxyphenoxy phenyl ethan-1-one, 4'-4-methoxyphenoxy acetophenone, 1-acetyl-4-4-methoxyphenoxy benzene, 1-4-4-methoxy-phenoxy-phenyl, maybridge1_003245, 4-acetyl-4'-methoxydiphenylether, 1-4-4-methoxy-phenoxy-phenyl-ethanone
PubChem CID 2777253
IUPAC Name 1-[4-(4-methoxyphenoxy)phenyl]ethanone
SMILES CC(=O)C1=CC=C(C=C1)OC2=CC=C(C=C2)OC

Specifications

Melting Point 59°C
Color White
Boiling Point 236°C
Quantity 1 g
Formula Weight 242.27
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4′-(4-Methoxyphenoxy)acetophenone