missing translation for 'onlineSavingsMsg'
Learn More

4-(4-Methoxybenzyloxy)-1,3-butanediol 95.0+%, TCI America™

Catalog No. M20645G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M20645G 5 g
M20641G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M20645G Supplier TCI America Supplier No. M20645G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1820740-81-5
Molecular Formula C12H18O4
Molecular Weight (g/mol) 226.272
InChI Key GCVOAHFIFQYCKH-UHFFFAOYSA-N
PubChem CID 44629983
IUPAC Name 4-[(4-methoxyphenyl)methoxy]butane-1,3-diol
SMILES COC1=CC=C(C=C1)COCC(CCO)O

Specifications

Color Colorless
Quantity 5 g
Formula Weight 226.27
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-(4-Methoxybenzyloxy)-1,3-butanediol