missing translation for 'onlineSavingsMsg'
Learn More

4,4-MDI, MilliporeSigma™ Supelco™

Catalog No. 111019613
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111019613 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111019613 Supplier Merck Emd Millipore Supplier No. 33428100MGR

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

4,4′-MDI can be extensively used to make urethane elastomers and urethane coatings.

Environmental

Specifications

CAS 101-68-8
Density 1.18 g/mL (at 25°C)
Flash Point 113°C - Closed Cup
Physical Form Neat
Linear Formula CH2(C6H4 NCO)2
MDL Number MFCD00036131
Melting Point 42°C to 45°C
Boiling Point 200°C (5 mmHg)
Synonym 4,4 ′-Methylenebis(phenyl isocyanate); 4,4 ′-MDI; Bis(4-isocyanatophenyl)methane
Type Analytical Standard
Quantity 100 mg
Formula Weight 250.25
Grade Analytical Standard
Show More Show Less