missing translation for 'onlineSavingsMsg'
Learn More

4-(4-Isopropylpiperazin-1-yl)phenylboronic acid pinacol ester, 97%, Thermo Scientific Chemicals

Catalog No. p-4813001
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC445080010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC445080010 Supplier Thermo Scientific Chemicals Supplier No. 445080010
Only null left

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1073354-18-3
Molecular Formula C19H31BN2O2
Molecular Weight (g/mol) 330.28
MDL Number MFCD06795656
InChI Key CSORKGLMGUQQOY-UHFFFAOYSA-N
Synonym 4-4-isopropylpiperizinyl phenylboronic acid, pinacol ester, 1-isopropyl-4-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl piperazine, 4-4-isopropylpiperizinyl phenylboronic acid pinacol ester, 4-4-isopropylpiperizinyl phenylboronicacid,pinacolester, 4-4-isopropylpiperazinyl phenylboronic acid, pinacol ester, 4-4-isopropylpiperizinyl benzeneboronic acid, pinacol ester, 4-4-isopropylpiperazin-1-yl phenylboronic acid pinacol ester, 1-propan-2-yl-4-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl piperazine, 1-isopropyl-4-4-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl piperazine
PubChem CID 17750254
IUPAC Name 1-propan-2-yl-4-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]piperazine
SMILES CC(C)N1CCN(CC1)C1=CC=C(C=C1)B1OC(C)(C)C(C)(C)O1

Specifications

Melting Point 139°C to 141°C
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 330.28
Percent Purity 97%
Chemical Name or Material 4-(4-Isopropylpiperazin-1-yl)phenylboronic acid pinacol ester
Hazard Category GHS Signal Word: Warning

RUO – Research Use Only