missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Isopropylidenediphenol Dimethacrylate 98.0+%, TCI America™

Catalog No. I038925G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I038925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. I038925G Supplier TCI America Supplier No. I038925G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 3253-39-2
Molecular Formula C23H24O4
Molecular Weight (g/mol) 364.44
MDL Number MFCD00008585
InChI Key QUZSUMLPWDHKCJ-UHFFFAOYSA-N
Synonym bisphenol a dimethacrylate, 2,2-di 4-methacryloxyphenyl propane, 4,4'-isopropylidenediphenol dimethacrylate, 4,4'-isopropylidenediphenyl dimethacrylate, 2-propenoic acid, 2-methyl-, 1-methylethylidene di-4,1-phenylene ester, propane-2,2-diylbis 4,1-phenylene bis 2-methylacrylate, 4-2-4-2-methylprop-2-enoyloxy phenyl propan-2-yl phenyl 2-methylprop-2-enoate, bis-dma, 2-propenoic acid, 2-methyl-, 1,1'-1-methylethylidene di-4,1-phenylene ester
PubChem CID 76739
ChEBI CHEBI:34579
IUPAC Name 4-(2-{4-[(2-methylprop-2-enoyl)oxy]phenyl}propan-2-yl)phenyl 2-methylprop-2-enoate
SMILES CC(=C)C(=O)OC1=CC=C(C=C1)C(C)(C)C1=CC=C(OC(=O)C(C)=C)C=C1

Specifications

Melting Point 75°C
Color White
Quantity 25 g
Formula Weight 364.44
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4,4′-Isopropylidenediphenol Dimethacrylate