missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Isopropylidenediphenol, 97%

Catalog No. AC158241000
Change view
Click to view available options
Quantity:
100 g
500 g
2 kg
5 kg
Packaging:
Plastic bottle
Plastic drum
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC158241000 100 g Plastic bottle
AC158245000 500 g Plastic bottle
AC158240020 2 kg Plastic bottle
AC158240050 5 kg Plastic drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC158241000 Supplier Thermo Scientific Chemicals Supplier No. 158241000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 80-05-7 | C15H16O2 | 228.29 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 80-05-7
Molecular Formula C15H16O2
Molecular Weight (g/mol) 228.29
MDL Number MFCD00002366
InChI Key IISBACLAFKSPIT-UHFFFAOYSA-N
Synonym bisphenol a, 2,2-bis 4-hydroxyphenyl propane, 4,4'-isopropylidenediphenol, diphenylolpropane, bisphenol, diano, 4,4'-bisphenol a, 4,4'-propane-2,2-diyl diphenol, biphenol a, dian
PubChem CID 6623
ChEBI CHEBI:33216
IUPAC Name 4-[2-(4-hydroxyphenyl)propan-2-yl]phenol
SMILES CC(C)(C1=CC=C(O)C=C1)C1=CC=C(O)C=C1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 155.0°C to 158.0°C
Color Yellow to Orange
Boiling Point 220.0°C (4.0 mmHg)
Flash Point 227°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Plastic bottle
Linear Formula (CH3)2C(C6H4OH)2
Quantity 100 g
Beilstein 06,1011
Merck Index 15,1298
Solubility Information Solubility in water: practically insoluble. Other solubilities: soluble in dilute alkalies, soluble in alcohol, slightly soluble in ccl4
Formula Weight 228.29
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 4,4′-Isopropylidenediphenol, 97%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes serious eye damage.
May cause an allergic skin reaction.
May cause respiratory irritation.
May damage fertility.
Toxic to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a
EINECSNumber 201-245-8
RTECSNumber SL6300000
TSCA TSCA

RUO – Research Use Only