missing translation for 'onlineSavingsMsg'
Learn More

4-(4-Hydroxyphenyl)thiomorpholine 1,1-Dioxide 98.0+%, TCI America™

Catalog No. H10635G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H10635G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H10635G Supplier TCI America Supplier No. H10635G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 103661-13-8
Molecular Formula C10H13NO3S
Molecular Weight (g/mol) 227.28
MDL Number MFCD06797109
InChI Key NSHOLIWPDBJMPT-UHFFFAOYSA-N
Synonym 4-(1,1-Dioxothiomorpholino)phenol, 4-Thiomorpholinophenol 1′C,1′C-Dioxide
PubChem CID 21845223
IUPAC Name 4-(4-hydroxyphenyl)-1λ⁶-thiomorpholine-1,1-dione
SMILES OC1=CC=C(C=C1)N1CCS(=O)(=O)CC1

Specifications

Melting Point 156°C
Color White
Quantity 5 g
Formula Weight 227.28
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-(4-Hydroxyphenyl)thiomorpholine 1,1-Dioxide