missing translation for 'onlineSavingsMsg'
Learn More

4-(4-Hydroxyphenyl)cyclohexanone 95.0+%, TCI America™

Catalog No. p-7130976
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H13165G 5 g
H131625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H13165G Supplier TCI America Supplier No. H13165G
Only null left

Chemical Identifiers

CAS 105640-07-1
Molecular Formula C12H14O2
Molecular Weight (g/mol) 190.24
MDL Number MFCD00210693
InChI Key SLJYPZJZQIHNGU-UHFFFAOYSA-N
Synonym 4-4-hydroxyphenyl cyclohexanone, 4-4-hydroxyphenyl cyclohexan-1-one, 4-4-hydroxyphenyl-cyclohexanone, cyclohexanone, 4-4-hydroxyphenyl, 4-4-hydroxyphenyl-1-cyclohexanone, acmc-1cukv, 4-4-oxocyclohexyl phenol, ksc495c5l, 4-4'-hydroxyphenyl-cyclohexanone
PubChem CID 4598916
IUPAC Name 4-(4-hydroxyphenyl)cyclohexan-1-one
SMILES OC1=CC=C(C=C1)C1CCC(=O)CC1

Specifications

Melting Point 169°C
Color White
Quantity 5 g
Formula Weight 190.24
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-(4-Hydroxyphenyl)cyclohexanone