missing translation for 'onlineSavingsMsg'
Learn More

4-(4-Hydroxyphenoxy)benzoic Acid 99.0+%, TCI America™

Catalog No. H06421G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H06421G 1 g
H06425G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H06421G Supplier TCI America Supplier No. H06421G

Chemical Identifiers

CAS 500-76-5
Molecular Formula C13H10O4
Molecular Weight (g/mol) 230.219
MDL Number MFCD00059645
InChI Key KLXPCYHWTLAVLN-UHFFFAOYSA-N
PubChem CID 615542
IUPAC Name 4-(4-hydroxyphenoxy)benzoic acid
SMILES C1=CC(=CC=C1C(=O)O)OC2=CC=C(C=C2)O

Specifications

Melting Point 196°C
Quantity 1 g
Formula Weight 230.22
Percent Purity ≥99.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-(4-Hydroxyphenoxy)benzoic Acid