missing translation for 'onlineSavingsMsg'
Learn More

4-(4-Hydroxybutyl)thiomorpholine 1,1-Dioxide 98.0+%, TCI America™

Catalog No. H11425G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H11425G 5 g
H114225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H11425G Supplier TCI America Supplier No. H11425G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 59801-41-1
Molecular Formula C8H17NO3S
Molecular Weight (g/mol) 207.29
MDL Number MFCD09038525
InChI Key STKAHXPYHDVHGO-UHFFFAOYSA-N
Synonym 4-(1,1-Dioxothiomorpholino)-1-butanol
PubChem CID 12324072
IUPAC Name 4-(4-hydroxybutyl)-1λ⁶-thiomorpholine-1,1-dione
SMILES OCCCCN1CCS(=O)(=O)CC1

Specifications

Melting Point 65°C
Quantity 5 g
Formula Weight 207.29
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-(4-Hydroxybutyl)thiomorpholine 1,1-Dioxide
TSCA No
Recommended Storage Refrigerator