accessibility menu, dialog, popup

UserName

SOONOH

CAS RN 59801-41-1

IUPAC Name: 4-(4-hydroxybutyl)-1λ⁶-thiomorpholine-1,1-dione
Synonyms: 4-(4-hydroxybutyl)-1λ⁶-thiomorpholine-1,1-dione
Molecular Weight (g/mol): 207.29
Molecular Formula: C8H17NO3S
InChi Key: STKAHXPYHDVHGO-UHFFFAOYSA-N
SMILES: OCCCCN1CCS(=O)(=O)CC1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results