missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4,4'-(Hexafluoroisopropylidene)diphthalic Anhydride 98.0+%, TCI America™
Supplier: TCI America H077125G
Specifications
| 4,4′-(Hexafluoroisopropylidene)diphthalic Anhydride | |
| 246°C | |
| 3 (10g/L H2O soln. 20°C) | |
| MFCD00039143 | |
| 3261 | |
| QHHKLPCQTTWFSS-UHFFFAOYSA-N | |
| 5-[2-(1,3-dioxo-2-benzofuran-5-yl)-1,1,1,3,3,3-hexafluoropropan-2-yl]-2-benzofuran-1,3-dione | |
| 70677 | |
| ≥98.0% (GC,T) |
| 1107-00-2 | |
| White-Yellow | |
| C19H6F6O6 | |
| 25 g | |
| 4,4'-hexafluoroisopropylidene diphthalic anhydride, 4,4'-hexafluoroisopropylidene diphthalicanhydride, 5,5'-perfluoropropane-2,2-diyl bis isobenzofuran-1,3-dione, 2,2-bis 3,4-anhydrodicarboxyphenyl hexafluoropropane, 4,4'-hexafluoroisopropylidene bis phthalic anhydride, 6fda, 1,3-isobenzofurandione, 5,5'-2,2,2-trifluoro-1-trifluoromethyl ethylidene bis, 2,2-bis 3,4-dicarboxyphenyl-1,1,1,3,3,3-hexafluoropropane dianhydride, 5-2-1,3-dioxo-2-benzofuran-5-yl-1,1,1,3,3,3-hexafluoropropan-2-yl-2-benzofuran-1,3-dione, 2,2'-bis-3,4-dicarboxyphenyl hexafluoropropane dianhydride | |
| C1=CC2=C(C=C1C(C3=CC4=C(C=C3)C(=O)OC4=O)(C(F)(F)F)C(F)(F)F)C(=O)OC2=O | |
| 444.241 | |
| 444.24 | |
| Crystalline Powder |
Chemical Identifiers
| 1107-00-2 | |
| 444.241 | |
| QHHKLPCQTTWFSS-UHFFFAOYSA-N | |
| 70677 | |
| C1=CC2=C(C=C1C(C3=CC4=C(C=C3)C(=O)OC4=O)(C(F)(F)F)C(F)(F)F)C(=O)OC2=O |
| C19H6F6O6 | |
| MFCD00039143 | |
| 4,4'-hexafluoroisopropylidene diphthalic anhydride, 4,4'-hexafluoroisopropylidene diphthalicanhydride, 5,5'-perfluoropropane-2,2-diyl bis isobenzofuran-1,3-dione, 2,2-bis 3,4-anhydrodicarboxyphenyl hexafluoropropane, 4,4'-hexafluoroisopropylidene bis phthalic anhydride, 6fda, 1,3-isobenzofurandione, 5,5'-2,2,2-trifluoro-1-trifluoromethyl ethylidene bis, 2,2-bis 3,4-dicarboxyphenyl-1,1,1,3,3,3-hexafluoropropane dianhydride, 5-2-1,3-dioxo-2-benzofuran-5-yl-1,1,1,3,3,3-hexafluoropropan-2-yl-2-benzofuran-1,3-dione, 2,2'-bis-3,4-dicarboxyphenyl hexafluoropropane dianhydride | |
| 5-[2-(1,3-dioxo-2-benzofuran-5-yl)-1,1,1,3,3,3-hexafluoropropan-2-yl]-2-benzofuran-1,3-dione |
Safety and Handling
EINECSNumber : (5)-6595
TSCA : Yes