accessibility menu, dialog, popup

UserName

FFFOOOOOOFFF

CAS RN 1107-00-2

IUPAC Name: 5-[2-(1,3-dioxo-1,3-dihydro-2-benzofuran-5-yl)-1,1,1,3,3,3-hexafluoropropan-2-yl]-1,3-dihydro-2-benzofuran-1,3-dione
Synonyms: 5-[2-(1,3-dioxo-2-benzofuran-5-yl)-1,1,1,3,3,3-hexafluoropropan-2-yl]-2-benzofuran-1,3-dione
Molecular Weight (g/mol): 444.24
Molecular Formula: C19H6F6O6
InChi Key: QHHKLPCQTTWFSS-UHFFFAOYSA-N
SMILES: FC(F)(F)C(C1=CC=C2C(=O)OC(=O)C2=C1)(C1=CC=C2C(=O)OC(=O)C2=C1)C(F)(F)F
Percent Purity 
  • (3)
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)
  • (3)
Physical Form 
  • (5)
Grade 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (5)
  • (3)
  • (3)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (3)
  • (5)
  • (4)