missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4,4'-(Hexafluoroisopropylidene)diphthalic Anhydride 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 246°C |
| Color | White-Yellow |
| pH | 3 (10g/L H2O soln. 20°C) |
| Quantity | 100 g |
| UN Number | 3261 |
| Formula Weight | 444.24 |
| Percent Purity | ≥98.0% (GC,T) |
| Grade | Not Available |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 4,4′-(Hexafluoroisopropylidene)diphthalic Anhydride |
Safety and Handling
| EINECSNumber | (5)-6595 |
| TSCA | Yes |