missing translation for 'onlineSavingsMsg'
Learn More

4,4'-(Hexafluoroisopropylidene)diphthalic anhydride, 99%

Catalog No. p-7046742
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1615006 5 g
AAL1615014 25 g
AAL1615022 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL1615006 Supplier Thermo Scientific Chemicals Supplier No. L1615006
Only null left

CAS: 1107-00-2 | C19H6F6O6 | 444.241 g/mol

4,4'-(Hexafluoroisopropylidene) diphthalic anhydride is involved in the synthesis of polyamides derived from terphenylenes, which is used for gas separation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4,4′-(Hexafluoroisopropylidene) diphthalic anhydride is involved in the synthesis of polyamides derived from terphenylenes, which is used for gas separation.

Solubility
Miscible with water.

Notes
Moisture sensitive. Incompatible with strong oxidizing agents and strong acids.

Chemical Identifiers

CAS 1107-00-2
Molecular Formula C19H6F6O6
Molecular Weight (g/mol) 444.241
MDL Number MFCD00039143
InChI Key QHHKLPCQTTWFSS-UHFFFAOYSA-N
Synonym 4,4'-hexafluoroisopropylidene diphthalic anhydride, 4,4'-hexafluoroisopropylidene diphthalicanhydride, 5,5'-perfluoropropane-2,2-diyl bis isobenzofuran-1,3-dione, 2,2-bis 3,4-anhydrodicarboxyphenyl hexafluoropropane, 4,4'-hexafluoroisopropylidene bis phthalic anhydride, 6fda, 1,3-isobenzofurandione, 5,5'-2,2,2-trifluoro-1-trifluoromethyl ethylidene bis, 2,2-bis 3,4-dicarboxyphenyl-1,1,1,3,3,3-hexafluoropropane dianhydride, 5-2-1,3-dioxo-2-benzofuran-5-yl-1,1,1,3,3,3-hexafluoropropan-2-yl-2-benzofuran-1,3-dione, 2,2'-bis-3,4-dicarboxyphenyl hexafluoropropane dianhydride
PubChem CID 70677
IUPAC Name 5-[2-(1,3-dioxo-2-benzofuran-5-yl)-1,1,1,3,3,3-hexafluoropropan-2-yl]-2-benzofuran-1,3-dione
SMILES C1=CC2=C(C=C1C(C3=CC4=C(C=C3)C(=O)OC4=O)(C(F)(F)F)C(F)(F)F)C(=O)OC2=O

Specifications

Melting Point 244°C to 247°C
Quantity 5 g
Beilstein 7057916
Sensitivity Moisture sensitive
Solubility Information Miscible with water.
Formula Weight 444.25
Percent Purity 99%
Chemical Name or Material 4,4'-(Hexafluoroisopropylidene)diphthalic anhydride
Hazard Category H314-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
EINECSNumber 214-170-0
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only