missing translation for 'onlineSavingsMsg'
Learn More

4,4'-(Hexafluoroisopropylidene)diphenol, 98%

CAS: 1478-61-1 | C15H10F6O2 | 336.23 g/mol

Supplier:  Thermo Scientific Chemicals A1837014

 View more versions of this product

Catalog No. AAA1837014


Only null left
Explore available promotions

Description

Description

4,4'-(Hexafluoroisopropylidene)diphenol is used to cross-link and cure fluoroelastomers. is also used as a monomer in the synthesis of specialty polymers. Polymers containing Bisphenol AF are useful in high temperature composites, in electronic materials and other specialty applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4,4′-(Hexafluoroisopropylidene)diphenol is used to cross-link and cure fluoroelastomers. is also used as a monomer in the synthesis of specialty polymers. Polymers containing Bisphenol AF are useful in high temperature composites, in electronic materials and other specialty applications.

Solubility
Insoluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Specifications

Specifications

4,4'-(Hexafluoroisopropylidene)diphenol
158°C to 162°C
C15H10F6O2
25 g
bisphenol af, 4,4'-hexafluoroisopropylidene diphenol, 2,2-bis 4-hydroxyphenyl hexafluoropropane, hexafluorobisphenol a, biphenol af, 4,4'-perfluoropropane-2,2-diyl diphenol, hexafluoroisopropylidenebis 4-hydroxybenzene, hexafluorodiphenylolpropane, hexafluoroacetone bisphenol a, 2,2-bis 4-hydroxyphenyl perfluoropropane
ZFVMWEVVKGLCIJ-UHFFFAOYSA-N
4-[1,1,1,3,3,3-hexafluoro-2-(4-hydroxyphenyl)propan-2-yl]phenol
73864
336.24
1478-61-1
400°C
MFCD00000439
1891568
Insoluble in water.
OC1=CC=C(C=C1)C(C1=CC=C(O)C=C1)(C(F)(F)F)C(F)(F)F
336.23
CHEBI:72754
98%

Chemical Identifiers

1478-61-1
336.23
ZFVMWEVVKGLCIJ-UHFFFAOYSA-N
73864
4-[1,1,1,3,3,3-hexafluoro-2-(4-hydroxyphenyl)propan-2-yl]phenol
C15H10F6O2
MFCD00000439
bisphenol af, 4,4'-hexafluoroisopropylidene diphenol, 2,2-bis 4-hydroxyphenyl hexafluoropropane, hexafluorobisphenol a, biphenol af, 4,4'-perfluoropropane-2,2-diyl diphenol, hexafluoroisopropylidenebis 4-hydroxybenzene, hexafluorodiphenylolpropane, hexafluoroacetone bisphenol a, 2,2-bis 4-hydroxyphenyl perfluoropropane
CHEBI:72754
OC1=CC=C(C=C1)C(C1=CC=C(O)C=C1)(C(F)(F)F)C(F)(F)F
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 216-036-7

RTECSNumber : SN2780000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only