missing translation for 'onlineSavingsMsg'
Learn More

4,4'-(Hexafluoroisopropylidene)dianiline, 98%, Thermo Scientific Chemicals

Catalog No. AC310990010
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC310990010 1 g Glass bottle
AC310990050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC310990010 Supplier Thermo Scientific Chemicals Supplier No. 310990010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1095-78-9
Molecular Formula C15H12F6N2
Molecular Weight (g/mol) 334.27
MDL Number MFCD00039146
InChI Key BEKFRNOZJSYWKZ-UHFFFAOYSA-N
Synonym 2,2-bis 4-aminophenyl hexafluoropropane, 4,4'-perfluoropropane-2,2-diyl dianiline, 4,4'-hexafluoroisopropylidene dianiline, 4-2-4-aminophenyl-1,1,1,3,3,3-hexafluoropropan-2-yl aniline, benzenamine, 4,4'-2,2,2-trifluoro-1-trifluoromethyl ethylidene bis, acmc-1bon7, bekfrnozjsywkz-uhfffaoysa, 2,2-bis-4-aminophenyl hexafluoropropane, 4,4'-1,1,1,3,3,3-hexafluoropropane-2,2-diyl dianiline
PubChem CID 622236
IUPAC Name 4-[2-(4-aminophenyl)-1,1,1,3,3,3-hexafluoropropan-2-yl]aniline
SMILES C1=CC(=CC=C1C(C2=CC=C(C=C2)N)(C(F)(F)F)C(F)(F)F)N

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 195.0°C to 199.0°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula (CF3)2C(C6H4NH2)2
Quantity 1 g
Solubility Information Solubility in water: insoluble in water
Formula Weight 334.27
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 4, 4'-(Hexafluoroisopropylidene)dianiline
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only