missing translation for 'onlineSavingsMsg'
Learn More

4-(4-Heptylphenyl)benzoic Acid 95.0+%, TCI America™

Catalog No. H07095G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H07095G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H07095G Supplier TCI America Supplier No. H07095G
Only null left

Chemical Identifiers

CAS 58573-94-7
Molecular Formula C20H24O2
Molecular Weight (g/mol) 296.41
MDL Number MFCD00143231
InChI Key XVROSBHWACAQRL-UHFFFAOYSA-N
Synonym 4-Heptylbiphenyl-4′C-carboxylic Acid
PubChem CID 3086355
IUPAC Name 4-(4-heptylphenyl)benzoic acid
SMILES CCCCCCCC1=CC=C(C=C1)C2=CC=C(C=C2)C(=O)O

Specifications

Color White-Yellow
Quantity 5 g
Formula Weight 296.41
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-(4-Heptylphenyl)benzoic Acid