missing translation for 'onlineSavingsMsg'
Learn More

4-(4-Fluorophenylethynyl)phenol 98.0+%, TCI America™

Catalog No. F04231G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F04231G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F04231G Supplier TCI America Supplier No. F04231G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 197770-48-2
Molecular Formula C14H9FO
Molecular Weight (g/mol) 212.223
MDL Number MFCD02093472
InChI Key LIPHCUGEWJPCQB-UHFFFAOYSA-N
Synonym 1-(4-Fluorophenyl)-2-(4-hydroxyphenyl)acetylene
PubChem CID 2782365
IUPAC Name 4-[2-(4-fluorophenyl)ethynyl]phenol
SMILES C1=CC(=CC=C1C#CC2=CC=C(C=C2)F)O

Specifications

Melting Point 149°C
Quantity 1 g
Formula Weight 212.22
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-(4-Fluorophenylethynyl)phenol