missing translation for 'onlineSavingsMsg'
Learn More

4-[[(4-Fluorophenyl)imino]methyl]phenol 98.0+%, TCI America™

Catalog No. F09465G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F09465G 5 g
F094625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F09465G Supplier TCI America Supplier No. F09465G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 3382-63-6
Molecular Formula C13H10FNO
Molecular Weight (g/mol) 215.227
MDL Number MFCD00029739
InChI Key QVDWVRKNMDWNRU-UHFFFAOYSA-N
PubChem CID 5397240
IUPAC Name 4-[(4-fluoroanilino)methylidene]cyclohexa-2,5-dien-1-one
SMILES C1=CC(=O)C=CC1=CNC2=CC=C(C=C2)F

Specifications

Melting Point 181°C
Color Yellow
Quantity 5 g
Formula Weight 215.23
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-[[(4-Fluorophenyl)imino]methyl]phenol
EINECSNumber (3)-4534
TSCA No