accessibility menu, dialog, popup

UserName

OHNF

CAS RN 3382-63-6

IUPAC Name: 4-{[(4-fluorophenyl)amino]methylidene}cyclohexa-2,5-dien-1-one
Synonyms: 4-{[(4-fluorophenyl)amino]methylidene}cyclohexa-2,5-dien-1-one
Molecular Weight (g/mol): 215.23
Molecular Formula: C13H10FNO
InChi Key: QVDWVRKNMDWNRU-UHFFFAOYSA-N
SMILES: FC1=CC=C(NC=C2C=CC(=O)C=C2)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results