missing translation for 'onlineSavingsMsg'
Learn More

4-(4-Fluorophenoxy)benzonitrile 98.0+%, TCI America™

Catalog No. F05321G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F05321G 1 g
F05325G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F05321G Supplier TCI America Supplier No. F05321G
Only null left

Chemical Identifiers

CAS 215589-24-5
Molecular Formula C13H8FNO
Molecular Weight (g/mol) 213.211
InChI Key NXVPHQNXBDDVCT-UHFFFAOYSA-N
Synonym 4-4-fluorophenoxy benzonitrile, 4-4-fluorophenoxy benzenecarbonitrile, acmc-1chxs, 4-cyano-4'-fluorodiphenyl ether, benzonitrile,4-4-fluorophenoxy
PubChem CID 1490284
IUPAC Name 4-(4-fluorophenoxy)benzonitrile
SMILES C1=CC(=CC=C1C#N)OC2=CC=C(C=C2)F

Specifications

Melting Point 69°C
Color White
Quantity 1 g
Formula Weight 213.21
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-(4-Fluorophenoxy)benzonitrile