missing translation for 'onlineSavingsMsg'
Learn More

4-(4-Fluorophenoxy)benzaldehyde 98.0+%, TCI America™

Catalog No. F05375G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F05375G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F05375G Supplier TCI America Supplier No. F05375G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 137736-06-2
Molecular Formula C13H9FO2
Molecular Weight (g/mol) 216.21
MDL Number MFCD01631896
InChI Key YUPBWHURNLRZQL-UHFFFAOYSA-N
Synonym 4-4-fluorophenoxy benzaldehyde, 4-4'-fluorophenoxy benzaldehyde, 4-4-fluoro-phenoxy-benzaldehyde, benzaldehyde, 4-4-fluorophenoxy, pubchem23015, acmc-209ceq, yupbwhurnlrzql-uhfffaoysa, 4-4-fluorophenoxyl benzaldehyde, 4-fluoro-4'-formyldiphenyl ether
PubChem CID 3856802
IUPAC Name 4-(4-fluorophenoxy)benzaldehyde
SMILES FC1=CC=C(OC2=CC=C(C=O)C=C2)C=C1

Specifications

Melting Point 77°C
Color Yellow
Quantity 5 g
UN Number 3077
Formula Weight 216.21
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-(4-Fluorophenoxy)benzaldehyde