missing translation for 'onlineSavingsMsg'
Learn More

4-(4-Fluorophenoxy)benzaldehyde, 97%

Catalog No. p-7033352
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH50452MD 250 mg
AAH5045203 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH50452MD Supplier Thermo Scientific Chemicals Supplier No. H50452MD

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 137736-06-2 | C13H9FO2 | 216.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 137736-06-2
Molecular Formula C13H9FO2
Molecular Weight (g/mol) 216.21
MDL Number MFCD01631896
InChI Key YUPBWHURNLRZQL-UHFFFAOYSA-N
Synonym 4-4-fluorophenoxy benzaldehyde, 4-4'-fluorophenoxy benzaldehyde, 4-4-fluoro-phenoxy-benzaldehyde, benzaldehyde, 4-4-fluorophenoxy, pubchem23015, acmc-209ceq, yupbwhurnlrzql-uhfffaoysa, 4-4-fluorophenoxyl benzaldehyde, 4-fluoro-4'-formyldiphenyl ether
PubChem CID 3856802
IUPAC Name 4-(4-fluorophenoxy)benzaldehyde
SMILES FC1=CC=C(OC2=CC=C(C=O)C=C2)C=C1

Specifications

Quantity 250 mg
Formula Weight 216.21
Percent Purity 97%
Chemical Name or Material 4-(4-Fluorophenoxy)benzaldehyde
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only