missing translation for 'onlineSavingsMsg'
Learn More

4,4'-(Ethyne-1,2-diyl)diphthalic Anhydride 98.0+%, TCI America™

Supplier:  TCI America E116425G

 View more versions of this product

Catalog No. E116425G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

4,4′-(Ethyne-1,2-diyl)diphthalic Anhydride
350°C
25 g
XGZRRDYHYZLYIJ-UHFFFAOYSA-N
5-[2-(1,3-dioxo-2-benzofuran-5-yl)ethynyl]-2-benzofuran-1,3-dione
58986478
≥98.0% (HPLC,T)
129808-00-0
C18H6O6
5,5′-(Ethyne-1,2-diyl)bis(isobenzofuran-1,3-dione)
C1=CC2=C(C=C1C#CC3=CC4=C(C=C3)C(=O)OC4=O)C(=O)OC2=O
318.24
318.24
Crystalline Powder

Chemical Identifiers

129808-00-0
318.24
58986478
C1=CC2=C(C=C1C#CC3=CC4=C(C=C3)C(=O)OC4=O)C(=O)OC2=O
C18H6O6
XGZRRDYHYZLYIJ-UHFFFAOYSA-N
5-[2-(1,3-dioxo-2-benzofuran-5-yl)ethynyl]-2-benzofuran-1,3-dione
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents

Promotions

Promotions