accessibility menu, dialog, popup

UserName

CAS RN 129808-00-0

OOOOOO

CAS RN 129808-00-0

IUPAC Name: 5-[2-(1,3-dioxo-1,3-dihydro-2-benzofuran-5-yl)ethynyl]-1,3-dihydro-2-benzofuran-1,3-dione
Synonyms: 5-[2-(1,3-dioxo-2-benzofuran-5-yl)ethynyl]-2-benzofuran-1,3-dione
Molecular Weight (g/mol): 318.24
Molecular Formula: C18H6O6
InChi Key: XGZRRDYHYZLYIJ-UHFFFAOYSA-N
SMILES: O=C1OC(=O)C2=C1C=CC(=C2)C#CC1=CC2=C(C=C1)C(=O)OC2=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results