missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4,4'-Ethylenedianiline 97.0+%, TCI America™
Supplier: TCI America E034625G
Specifications
| 4,4′-Ethylenedianiline | |
| 134°C | |
| MFCD00007923 | |
| 4,4'-ethylenedianiline, 4,4'-diaminobibenzyl, 4-2-4-aminophenyl ethyl aniline, 4,4'-diaminodibenzyl, 1,2-bis p-aminophenyl ethane, benzenamine, 4,4'-1,2-ethanediyl bis, 4,4'-ethylenebis aniline, aniline, 4,4'-ethylenedi, .alpha.,.alpha.'-bi-p-toluidine, 4-2-4-aminophenyl ethyl phenylamine | |
| C1=CC(=CC=C1CCC2=CC=C(C=C2)N)N | |
| 212.296 | |
| 212.30 | |
| Crystalline Powder |
| 621-95-4 | |
| C14H16N2 | |
| 25 g | |
| UHNUHZHQLCGZDA-UHFFFAOYSA-N | |
| 4-[2-(4-aminophenyl)ethyl]aniline | |
| 12138 | |
| ≥97.0% (GC,T) |
Chemical Identifiers
| 621-95-4 | |
| 212.296 | |
| UHNUHZHQLCGZDA-UHFFFAOYSA-N | |
| 12138 | |
| C1=CC(=CC=C1CCC2=CC=C(C=C2)N)N |
| C14H16N2 | |
| MFCD00007923 | |
| 4,4'-ethylenedianiline, 4,4'-diaminobibenzyl, 4-2-4-aminophenyl ethyl aniline, 4,4'-diaminodibenzyl, 1,2-bis p-aminophenyl ethane, benzenamine, 4,4'-1,2-ethanediyl bis, 4,4'-ethylenebis aniline, aniline, 4,4'-ethylenedi, .alpha.,.alpha.'-bi-p-toluidine, 4-2-4-aminophenyl ethyl phenylamine | |
| 4-[2-(4-aminophenyl)ethyl]aniline |
Safety and Handling
RTECSNumber : BY0200000
TSCA : Yes