accessibility menu, dialog, popup

UserName

NH2NH2

CAS RN 621-95-4

IUPAC Name: 4-[2-(4-aminophenyl)ethyl]aniline
Synonyms: 4-[2-(4-aminophenyl)ethyl]aniline
Molecular Weight (g/mol): 212.3
Molecular Formula: C14H16N2
InChi Key: UHNUHZHQLCGZDA-UHFFFAOYSA-N
SMILES: NC1=CC=C(CCC2=CC=C(N)C=C2)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results