missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Dithiodipyridine, 98%

Catalog No. p-124257
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC162240010 1 g Glass bottle
AC162240050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC162240010 Supplier Thermo Scientific Chemicals Supplier No. 162240010
Only null left

CAS: 2645-22-9 | C10H8N2S2 | 220.32 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2645-22-9
Molecular Formula C10H8N2S2
Molecular Weight (g/mol) 220.32
MDL Number MFCD00006423
InChI Key UHBAPGWWRFVTFS-UHFFFAOYSA-N
Synonym 4,4'-dithiodipyridine, 4,4'-dipyridyl disulfide, 1,2-di pyridin-4-yl disulfane, aldrithiol-4, pyridine, 4,4'-dithiobis, 4,4'-dithiopyridine, di 4-pyridyl disulfide, aldrithiol 4, di 4-pyridinyl disulfide, 4,4'-dipyridine disulfide
PubChem CID 75846
ChEBI CHEBI:41814
IUPAC Name 4-(pyridin-4-yldisulfanyl)pyridine
SMILES C1=CN=CC=C1SSC2=CC=NC=C2

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 75°C to 79°C
Color Orange to White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula (C5H4N)S-S(C5H4N)
Quantity 1 g
Beilstein 21, II, 35
Formula Weight 220.32
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 4, 4'-Dithiodipyridine, 98%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Avoid breath
EINECSNumber 220-158-6
TSCA TSCA

RUO – Research Use Only