missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Dithiodimorpholine 98.0+%, TCI America™

Catalog No. d028225g
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D028225G 25 g
D0282500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D028225G Supplier TCI America Supplier No. D028225G
Only null left

Chemical Identifiers

CAS 103-34-4
Molecular Formula C8H16N2O2S2
Molecular Weight (g/mol) 236.348
MDL Number MFCD00023319
InChI Key HLBZWYXLQJQBKU-UHFFFAOYSA-N
PubChem CID 7648
IUPAC Name 4-(morpholin-4-yldisulfanyl)morpholine
SMILES C1COCCN1SSN2CCOCC2

Specifications

Melting Point 125°C
Color White-Yellow
Quantity 25 g
Synonym N,N′C-Dimorpholine Disulfide, N,N′C-Dithiobis(morpholine)
Formula Weight 236.35
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 4,4′-Dithiodimorpholine
EINECSNumber (5)-0892
RTECSNumber QE3325000
TSCA Yes
Recommended Storage Refrigerator