accessibility menu, dialog, popup

UserName

CAS RN 103-34-4

NOSSNO

CAS RN 103-34-4

IUPAC Name: 4-(morpholin-4-yldisulfanyl)morpholine
Synonyms: morpholine, 4,4'-dithiobis-
Molecular Weight (g/mol): 236.35
Molecular Formula: C8H16N2O2S2
InChi Key: HLBZWYXLQJQBKU-UHFFFAOYSA-N
SMILES: C1CN(CCO1)SSN1CCOCC1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results