missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4,4'-Dipyridyl Disulfide 97.0+%, TCI America™
Supplier: TCI America D247725G
Specifications
| 4,4′-Dipyridyl Disulfide | |
| 75°C | |
| MFCD00006423 | |
| 4,4'-dithiodipyridine, 4,4'-dipyridyl disulfide, 1,2-di pyridin-4-yl disulfane, aldrithiol-4, pyridine, 4,4'-dithiobis, 4,4'-dithiopyridine, di 4-pyridyl disulfide, aldrithiol 4, di 4-pyridinyl disulfide, 4,4'-dipyridine disulfide | |
| C1=CN=CC=C1SSC2=CC=NC=C2 | |
| 220.308 | |
| CHEBI:41814 | |
| ≥97.0% (GC,T) |
| 2645-22-9 | |
| C10H8N2S2 | |
| 25 g | |
| UHBAPGWWRFVTFS-UHFFFAOYSA-N | |
| 4-(pyridin-4-yldisulfanyl)pyridine | |
| 75846 | |
| 220.31 | |
| Crystalline Powder |
Chemical Identifiers
| 2645-22-9 | |
| 220.308 | |
| UHBAPGWWRFVTFS-UHFFFAOYSA-N | |
| 75846 | |
| 4-(pyridin-4-yldisulfanyl)pyridine |
| C10H8N2S2 | |
| MFCD00006423 | |
| 4,4'-dithiodipyridine, 4,4'-dipyridyl disulfide, 1,2-di pyridin-4-yl disulfane, aldrithiol-4, pyridine, 4,4'-dithiobis, 4,4'-dithiopyridine, di 4-pyridyl disulfide, aldrithiol 4, di 4-pyridinyl disulfide, 4,4'-dipyridine disulfide | |
| CHEBI:41814 | |
| C1=CN=CC=C1SSC2=CC=NC=C2 |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator