missing translation for 'onlineSavingsMsg'
Learn More

4,4-Diphenylcyclohexanol 98.0+%, TCI America™

Catalog No. D49735G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D49735G 5 g
D49731G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D49735G Supplier TCI America Supplier No. D49735G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 42420-85-9
Molecular Formula C18H20O
Molecular Weight (g/mol) 252.357
MDL Number MFCD00087400
InChI Key UUQQLWRHPNEFIG-UHFFFAOYSA-N
PubChem CID 10966998
IUPAC Name 4,4-diphenylcyclohexan-1-ol
SMILES C1CC(CCC1O)(C2=CC=CC=C2)C3=CC=CC=C3

Specifications

Melting Point 143°C
Color White
Quantity 5 g
Formula Weight 252.36
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4,4-Diphenylcyclohexanol