missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Diphenoxybenzophenone, 98%

CAS: 14984-21-5 | C25H18O3 | 366.42 g/mol

Supplier:  Thermo Scientific Chemicals L1341906

 View more versions of this product

Catalog No. AAL1341906


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

4,4'-Diphenoxybenzophenone
147°C to 151°C
MFCD00077969
2165449
BSILAEQTGTZMIW-UHFFFAOYSA-N
bis(4-phenoxyphenyl)methanone
84743
98%
14984-21-5
C25H18O3
5 g
4,4'-diphenoxybenzophenone, bis 4-phenoxyphenyl methanone, methanone, bis 4-phenoxyphenyl, unii-2l24ex5o0x, benzophenone, 4,4'-diphenoxy, di4-phenoxyphenyl ketone, zlchem 703, acmc-1br23, 4,4/'-diphenoxybenzophenone, 4,4'-bis phenoxy benzophenone
O=C(C1=CC=C(OC2=CC=CC=C2)C=C1)C1=CC=C(OC2=CC=CC=C2)C=C1
366.42
366.42

Chemical Identifiers

14984-21-5
366.42
BSILAEQTGTZMIW-UHFFFAOYSA-N
84743
O=C(C1=CC=C(OC2=CC=CC=C2)C=C1)C1=CC=C(OC2=CC=CC=C2)C=C1
C25H18O3
MFCD00077969
4,4'-diphenoxybenzophenone, bis 4-phenoxyphenyl methanone, methanone, bis 4-phenoxyphenyl, unii-2l24ex5o0x, benzophenone, 4,4'-diphenoxy, di4-phenoxyphenyl ketone, zlchem 703, acmc-1br23, 4,4/'-diphenoxybenzophenone, 4,4'-bis phenoxy benzophenone
bis(4-phenoxyphenyl)methanone
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 403-390-4

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only