missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Diphenoxybenzophenone, 98%

Catalog No. AAL1341903
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1341903 1 g
AAL1341906 5 g
AAL1341914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL1341903 Supplier Thermo Scientific Chemicals Supplier No. L1341903
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 14984-21-5 | C25H18O3 | 366.42 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 14984-21-5
Molecular Formula C25H18O3
Molecular Weight (g/mol) 366.42
MDL Number MFCD00077969
InChI Key BSILAEQTGTZMIW-UHFFFAOYSA-N
Synonym 4,4'-diphenoxybenzophenone, bis 4-phenoxyphenyl methanone, methanone, bis 4-phenoxyphenyl, unii-2l24ex5o0x, benzophenone, 4,4'-diphenoxy, di4-phenoxyphenyl ketone, zlchem 703, acmc-1br23, 4,4/'-diphenoxybenzophenone, 4,4'-bis phenoxy benzophenone
PubChem CID 84743
IUPAC Name bis(4-phenoxyphenyl)methanone
SMILES O=C(C1=CC=C(OC2=CC=CC=C2)C=C1)C1=CC=C(OC2=CC=CC=C2)C=C1

Specifications

Melting Point 147°C to 151°C
Quantity 1 g
Beilstein 2165449
Formula Weight 366.42
Percent Purity 98%
Chemical Name or Material 4,4'-Diphenoxybenzophenone
EINECSNumber 403-390-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only