missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4,4-Dioxo-1,4-oxathiane 98.0+%, TCI America™
Supplier: TCI America D21085G
Specifications
| 4,4-Dioxo-1,4-oxathiane | |
| 133°C | |
| C4H8O3S | |
| 5 g | |
| WWRUZECKUVNAPB-UHFFFAOYSA-N | |
| 1,4λâ¶-oxathiane-4,4-dione | |
| 7878 | |
| ≥98.0% (GC) |
| 107-61-9 | |
| White | |
| MFCD00011683 | |
| 1,4-thioxane-1,1-dioxide, thioxane sulfone, p-thioxane sulfone, 1,4-oxathiane, 4,4-dioxide, oxathiane 4,4-dioxide, usaf do-38, 4,4-dioxo-1,4-oxathiane, 1,4-thioxane 1,1-dioxide, 1,4lambda6-oxathiane-4,4-dione, thioxanesulfone | |
| O=S1(=O)CCOCC1 | |
| 136.17 | |
| 136.17 | |
| Crystalline Powder |
Chemical Identifiers
| 107-61-9 | |
| 136.17 | |
| WWRUZECKUVNAPB-UHFFFAOYSA-N | |
| 7878 | |
| O=S1(=O)CCOCC1 |
| C4H8O3S | |
| MFCD00011683 | |
| 1,4-thioxane-1,1-dioxide, thioxane sulfone, p-thioxane sulfone, 1,4-oxathiane, 4,4-dioxide, oxathiane 4,4-dioxide, usaf do-38, 4,4-dioxo-1,4-oxathiane, 1,4-thioxane 1,1-dioxide, 1,4lambda6-oxathiane-4,4-dione, thioxanesulfone | |
| 1,4λâ¶-oxathiane-4,4-dione |
Safety and Handling
RTECSNumber : RP4375000
TSCA : Yes