missing translation for 'onlineSavingsMsg'
Learn More

4,4-Dioxo-1,4-oxathiane 98.0+%, TCI America™

Catalog No. p-7127111
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D21085G 5 g
D210825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D21085G Supplier TCI America Supplier No. D21085G

Chemical Identifiers

CAS 107-61-9
Molecular Formula C4H8O3S
Molecular Weight (g/mol) 136.17
MDL Number MFCD00011683
InChI Key WWRUZECKUVNAPB-UHFFFAOYSA-N
Synonym 1,4-thioxane-1,1-dioxide, thioxane sulfone, p-thioxane sulfone, 1,4-oxathiane, 4,4-dioxide, oxathiane 4,4-dioxide, usaf do-38, 4,4-dioxo-1,4-oxathiane, 1,4-thioxane 1,1-dioxide, 1,4lambda6-oxathiane-4,4-dione, thioxanesulfone
PubChem CID 7878
IUPAC Name 1,4λ⁶-oxathiane-4,4-dione
SMILES O=S1(=O)CCOCC1

Specifications

Melting Point 133°C
Color White
Quantity 5 g
Formula Weight 136.17
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4,4-Dioxo-1,4-oxathiane
RTECSNumber RP4375000
TSCA Yes