missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Dinonyl-2,2'-bipyridyl 98.0+%, TCI America™

Catalog No. p-7127995
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D39171G 1 g
D39175G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D39171G Supplier TCI America Supplier No. D39171G

Chemical Identifiers

CAS 142646-58-0
Molecular Formula C28H44N2
Molecular Weight (g/mol) 408.674
MDL Number MFCD00800915
InChI Key VHJFWJXYEWHCGD-UHFFFAOYSA-N
Synonym 4,4'-dinonyl-2,2'-bipyridine, 4,4'-dinonyl-2,2'-bipyridyl, 4-nonyl-2-4-nonylpyridin-2-yl pyridine, 2,2'-bipyridine, 4,4'-dinonyl, 4,4'-bis nonyl-2,2'-bipyridine, dnbpy, acmc-1cctu, bpdn01, 4,4-dinonyl-2,2-bipyridine, 4,4-dinoyl-2,2-dipyridyl
PubChem CID 4285174
IUPAC Name 4-nonyl-2-(4-nonylpyridin-2-yl)pyridine
SMILES CCCCCCCCCC1=CC(=NC=C1)C2=NC=CC(=C2)CCCCCCCCC

Specifications

Melting Point 63°C
Color White
Quantity 1 g
Formula Weight 408.67
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4,4′-Dinonyl-2,2′-bipyridyl