accessibility menu, dialog, popup

UserName

H3CNNCH3

CAS RN 142646-58-0

IUPAC Name: 4,4'-dinonyl-2,2'-bipyridine
Synonyms: 4,4'-dinonyl-2,2'-bipyridine
Molecular Weight (g/mol): 408.67
Molecular Formula: C28H44N2
InChi Key: VHJFWJXYEWHCGD-UHFFFAOYSA-N
SMILES: CCCCCCCCCC1=CC(=NC=C1)C1=NC=CC(CCCCCCCCC)=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)