missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4,4'-Dinitrodiphenylmethane 99.0+%, TCI America™
$167.76 - $501.43
Chemical Identifiers
| CAS | 1817-74-9 |
|---|---|
| Molecular Formula | C13H10N2O4 |
| Molecular Weight (g/mol) | 258.23 |
| MDL Number | MFCD00059184 |
| InChI Key | GLBZQZXDUTUCGK-UHFFFAOYSA-N |
| PubChem CID | 15753 |
| IUPAC Name | 1-nitro-4-[(4-nitrophenyl)methyl]benzene |
| SMILES | [O-][N+](=O)C1=CC=C(CC2=CC=C(C=C2)[N+]([O-])=O)C=C1 |
Chemical Identifiers
| 1817-74-9 | |
| 258.23 | |
| GLBZQZXDUTUCGK-UHFFFAOYSA-N | |
| 1-nitro-4-[(4-nitrophenyl)methyl]benzene |
| C13H10N2O4 | |
| MFCD00059184 | |
| 15753 | |
| [O-][N+](=O)C1=CC=C(CC2=CC=C(C=C2)[N+]([O-])=O)C=C1 |
Specifications
| 1817-74-9 | |
| Yellow | |
| MFCD00059184 | |
| GLBZQZXDUTUCGK-UHFFFAOYSA-N | |
| 1-nitro-4-[(4-nitrophenyl)methyl]benzene | |
| 15753 | |
| ≥99.0% (GC) | |
| 4,4′-Dinitrodiphenylmethane |
| 190°C | |
| C13H10N2O4 | |
| 1 g | |
| [O-][N+](=O)C1=CC=C(CC2=CC=C(C=C2)[N+]([O-])=O)C=C1 | |
| 258.23 | |
| 258.23 | |
| Crystalline Powder |
Safety and Handling
TSCA : Yes