missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Dinitrodiphenylmethane 99.0+%, TCI America™

Catalog No. D08331G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D08331G 1 g
D08335G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D08331G Supplier TCI America Supplier No. D08331G
Only null left

Chemical Identifiers

CAS 1817-74-9
Molecular Formula C13H10N2O4
Molecular Weight (g/mol) 258.23
MDL Number MFCD00059184
InChI Key GLBZQZXDUTUCGK-UHFFFAOYSA-N
PubChem CID 15753
IUPAC Name 1-nitro-4-[(4-nitrophenyl)methyl]benzene
SMILES [O-][N+](=O)C1=CC=C(CC2=CC=C(C=C2)[N+]([O-])=O)C=C1

Specifications

Melting Point 190°C
Color Yellow
Quantity 1 g
Formula Weight 258.23
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4,4′-Dinitrodiphenylmethane