missing translation for 'onlineSavingsMsg'
Learn More

4,4-Dimethylcyclohexanone, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H2585706

 View more versions of this product

Catalog No. AAH2585706


Only null left
Explore available promotions

Description

Description

4,4-Dimethylcyclohexanone is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4,4-Dimethylcyclohexanone is used as a pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible material is strong oxidizing agents.
Specifications

Specifications

4,4-Dimethylcyclohexanone
41°C to 43°C
C8H14O
5 g
Insoluble in water.
CC1(CCC(=O)CC1)C
126.199
126.2
4255-62-3
62°C (144°F)
MFCD00234965
4,4-dimethylcyclohexanone, cyclohexanone, 4,4-dimethyl, 4,4-dimethyl cyclohexanone, 4,4-dimethyl-cyclohexan-1-one, 4,4-dimethycyclohexanone, 4.4-dimethylcyclohexanone, acmc-209jq8, 4,4-dimethyl-cyclohexanone, ksc494a0f, 4,4-dimethyl-1-cyclohexanone
PXQMSTLNSHMSJB-UHFFFAOYSA-N
4,4-dimethylcyclohexan-1-one
138166
98%

Chemical Identifiers

4255-62-3
126.199
PXQMSTLNSHMSJB-UHFFFAOYSA-N
138166
CC1(CCC(=O)CC1)C
C8H14O
MFCD00234965
4,4-dimethylcyclohexanone, cyclohexanone, 4,4-dimethyl, 4,4-dimethyl cyclohexanone, 4,4-dimethyl-cyclohexan-1-one, 4,4-dimethycyclohexanone, 4.4-dimethylcyclohexanone, acmc-209jq8, 4,4-dimethyl-cyclohexanone, ksc494a0f, 4,4-dimethyl-1-cyclohexanone
4,4-dimethylcyclohexan-1-one
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

P210-P240-P241-P264b-P280-P305+P351+P338-P370+P378q

H228-H319-H500

DOTInformation : Hazard Class: 4.1; Packaging Group: II

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only